Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037757.1 | NP_001032846.1 | 11 | 2855 | ||
NM_026833.1 | NP_081109.1 | 11 | 2872 | |||
GenBank mRNA | AK171737.1 | - | 11 | 2790 | ||
AK172673.1 | - | 11 | 2790 |