Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181395.2 | NP_852060.2 | 17 | 2311 | ||
GenBank mRNA | AK122223.1 | - | 17 | 2081 | ||
AK142872.1 | - | 17 | 2311 | |||
BC044828.1 | - | 2 | 275 | |||
BC049858.1 | - | 2 | 275 | |||
BC112913.1 | - | 8 | 1108 | |||
BC150789.1 | - | 17 | 2306 | 9087700 |