Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027357.2 | NP_081633.1 | 9, 10 | 1250 | ||
GenBank mRNA | AK087947.1 | - | 9, 10 | 1201 | ||
AK153386.1 | - | 9, 10 | 1251 | |||
AK159123.1 | - | 9, 10 | 1250 | |||
AK159750.1 | - | 9, 10 | 1229 | |||
AK215835.1 | - | 1, 2 | 148 | |||
BC022955.1 | - | 9, 10 | 1140 | |||
BC025802.1 | - | 9, 10 | 1154 | |||
BC029319.1 | - | 9, 10 | 1187 | |||
BC031587.1 | - | 9, 10 | 1181 | |||
BC032068.1 | - | 9, 10 | 1181 | |||
BC039937.1 | - | 9, 10 | 1227 | |||
BC089028.1 | - | 9, 10 | 1197 | |||
BC138526.1 | - | 9, 10 | 1213 | 8861546 | ||
BC138527.1 | - | 9, 10 | 1213 | 8861547 |