Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033163.3 | NP_001028335.1 | 3 | 683 | ||
GenBank mRNA | AK010021.1 | - | 3 | 670 | ||
AK132483.1 | - | 3 | 684 | |||
BC145797.1 | - | 3 | 748 | 40131100 |