Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700049G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028538.1 | NP_082814.1 | 4 | 4012 | ||
GenBank mRNA | AK137340.1 | - | 4 | 4012 |
Gene Symbol: Zfp780b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081021.1 | NP_001074490.1 | 6 | 5068 | ||
GenBank mRNA | AK137196.1 | - | 1 | 2053 | ||
BC094325.1 | - | 6 | 799 |