Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppapdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028922.3 | NP_083198.1 | 1 | 1178 | ||
GenBank mRNA | AK016572.1 | - | 1 | 1179 | ||
AK041581.1 | - | 1 | 1161 | |||
AK149181.1 | - | 1 | 945 | |||
BC052412.1 | - | 1 | 1145 | 5720146 |