Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 5 | 911 | ||
NM_001040195.1 | NP_001035285.1 | 4 | 876 | |||
NM_001040196.1 | NP_001035286.1 | 6 | 1008 | |||
NM_001040197.1 | NP_001035287.1 | 6 | 1029 | |||
NM_020350.4 | NP_065083.3 | 5 | 932 | |||
XM_011541799.2 | XP_011540101.2 | 6 | 925 | |||
XM_011541800.2 | XP_011540102.1 | 6 | 946 | |||
XM_011541802.1 | XP_011540104.1 | 7 | 1022 | |||
GenBank mRNA | AF312374.1 | - | 5 | 863 | ||
AK290042.1 | - | 5 | 875 | |||
BC011946.1 | - | 5 | 817 | |||
BC017328.2 | - | 5 | 849 | 4546585 | ||
BQ672962.1 | - | 1 | 416 |