Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 11, 12 | 2465 | ||
GenBank mRNA | AK041726.1 | - | 11, 12 | 2415 | ||
AK051639.1 | - | 11, 12 | 2428 | |||
AK054492.1 | - | 11, 12 | 2415 | |||
AK137597.1 | - | 11, 12 | 2470 | |||
AK157802.1 | - | 10, 11 | 4022 | |||
BC158103.1 | - | 11, 12 | 2429 | 9088180 |