Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 5 | 441 | ||
GenBank mRNA | AK122507.1 | - | 4 | 1932 | ||
AK154554.1 | - | 5 | 437 | |||
BQ832419.1 | - | 1 | 46 | |||
CD539539.1 | - | 5 | 368 |
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166076.1 | - | 407 | |||
BC147312.1 | - | 421 | 9056375 | |||
BC147313.1 | - | 421 | 9056366 |