Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 6, 7 | 609 | ||
GenBank mRNA | AK122507.1 | - | 5, 6 | 2100 | ||
AK154554.1 | - | 6, 7 | 605 | |||
BQ832419.1 | - | 2, 3 | 214 | |||
CD539539.1 | - | 6, 7 | 536 |
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166076.1 | - | 575 | |||
BC147312.1 | - | 589 | 9056375 | |||
BC147313.1 | - | 589 | 9056366 |