Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026876.3 | NP_081152.2 | 2, 3 | 568 | ||
GenBank mRNA | AK088077.1 | - | 2, 3 | 569 | ||
AK146088.1 | - | 2, 3 | 569 | |||
AK148524.1 | - | 2, 3 | 675 | |||
BC061491.1 | - | 2, 3 | 574 | |||
BC064024.1 | - | 2, 3 | 338 | |||
BC110425.1 | - | 2, 3 | 551 | 5369595 | ||
BC132299.1 | - | 2, 3 | 426 | 40130576 |