Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026876.3 | NP_081152.2 | 3 | 606 | ||
GenBank mRNA | AK088077.1 | - | 3 | 607 | ||
AK140206.1 | - | 2 | 2443 | |||
AK146088.1 | - | 3 | 607 | |||
AK148524.1 | - | 3 | 713 | |||
BC061491.1 | - | 3 | 612 | |||
BC064024.1 | - | 3 | 376 | |||
BC110425.1 | - | 3 | 589 | 5369595 | ||
BC132299.1 | - | 3 | 464 | 40130576 |