Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026876.3 | NP_081152.2 | 12 | 1940 | ||
GenBank mRNA | AK004493.1 | - | 4 | 433 | ||
AK146088.1 | - | 12 | 1941 | |||
BC022682.1 | - | 7 | 976 | |||
BC037478.1 | - | 8 | 1120 | |||
BC064024.1 | - | 12 | 1738 | |||
BC110425.1 | - | 12 | 1923 | 5369595 | ||
BC132299.1 | - | 12 | 1798 | 40130576 |