Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933402N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177510.1 | NP_001170981.1 | 3 | 376 | ||
GenBank mRNA | AK077088.1 | - | 441 | |||
BC133658.1 | - | 3 | 376 | 40050211 |
Gene Symbol: Gm5861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030042.1 | - | 357 |
Gene Symbol: Gm6455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048648.1 | - | 3 | 329 |
Gene Symbol: Gm6460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037919.3 | NP_001033008.1 | 3 | 376 | ||
GenBank mRNA | BC110565.1 | - | 3 | 376 | 40050213 |