Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037933.2 | NP_001033022.1 | 1, 2 | 84 | ||
GenBank mRNA | BC132568.1 | - | 1, 2 | 84 | 40130845 | |
BC132570.1 | - | 1, 2 | 84 | 40130847 | ||
DQ012034.1 | - | 1, 2 | 56 |