Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005568.4 | NP_005559.2 | 1 | 865 | ||
GenBank mRNA | AK298253.1 | - | 1 | 508 | ||
BC020470.1 | - | 1 | 865 | |||
BC101674.1 | - | 1 | 234 | 8069180 | ||
BC111936.1 | - | 1 | 234 | 8327404 | ||
U14755.1 | - | 1 | 574 |