Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006943.1 | NP_001006944.1 | 4 | 964 | ||
XM_011540969.1 | XP_011539271.1 | 6 | 1127 | |||
XM_011540970.1 | XP_011539272.1 | 4 | 861 | |||
XM_011540972.1 | XP_011539274.1 | 5 | 1028 | |||
XM_011540973.1 | XP_011539275.1 | 3 | 763 | |||
GenBank mRNA | AB040892.1 | - | 2 | 385 | ||
BC038796.1 | - | 4 | 894 | |||
BC072417.1 | - | 4 | 964 |