Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000570.4 | NP_000561.3 | 6 | 1701 | ||
NM_001244753.1 | NP_001231682.1 | 5 | 1947 | |||
NM_001271035.1 | NP_001257964.1 | 5 | 1944 | |||
NM_001271036.1 | NP_001257965.1 | 5 | 1813 | |||
NM_001271037.1 | NP_001257966.1 | 4 | 1555 | |||
GenBank mRNA | AJ581669.1 | - | 5 | 1613 |
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 6 | 1731 | ||
NM_001127595.1 | NP_001121067.1 | 6 | 1713 | |||
NM_001127596.1 | NP_001121068.1 | 6 | 1710 | |||
XM_011509293.2 | XP_011507595.2 | 4 | 1362 | |||
GenBank mRNA | AK223268.1 | - | 5 | 1592 | ||
AK223295.1 | - | 5 | 1603 | |||
BC033678.1 | - | 5 | 1599 | 5180561 | ||
BC036723.1 | - | 5 | 1868 | 5206097 |