Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097579.1 | NP_001091048.1 | 3 | 1325 | ||
NM_005300.3 | NP_005291.1 | 3 | 1249 | |||
XM_005272597.3 | XP_005272654.1 | 3 | 3752 | |||
GenBank mRNA | AA130392.1 | - | 1 | 197 | ||
AF039686.1 | - | 1 | 1119 | |||
AK027780.1 | - | 3 | 1325 | |||
AK074627.1 | - | 3 | 1471 | |||
AK075217.1 | - | 3 | 1240 | |||
AK122945.1 | - | 1 | 2680 | |||
BC020678.1 | - | 3 | 1216 | 4770479 | ||
DQ106398.1 | - | 3 | 1316 | |||
DQ106399.1 | - | 5 | 1287 | |||
DQ106400.1 | - | 5 | 1333 | |||
DQ106401.1 | - | 3 | 1175 | |||
DQ106402.1 | - | 4 | 1177 |
Set Membership: |
G-protein coupled receptor |