Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242758.1 | NP_001229687.1 | 8 | 1463 | ||
NM_002116.7 | NP_002107.3 | 8 | 1463 | |||
GenBank mRNA | AB209117.1 | - | 7 | 1546 | ||
AK130165.1 | - | 8 | 1418 | |||
AK304472.1 | - | 8 | 1357 | |||
BC003069.2 | - | 8 | 1380 | 3510139 | ||
BC019236.2 | - | 8 | 1384 | 2821717 | ||
BE875389.1 | - | 3 | 376 | |||
M11887.1 | - | 7 | 1109 | |||
M27971.1 | - | 5 | 635 | |||
X02457.1 | - | 7 | 1265 | |||
X13111.1 | - | 8 | 1379 |