Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166588.2 | NP_001160060.1 | 7 | 997 | ||
NM_001261418.1 | NP_001248347.1 | 7 | 997 | |||
NM_020365.4 | NP_065098.1 | 7 | 997 | |||
XM_017002745.1 | XP_016858234.1 | 7 | 885 | |||
XM_017002746.1 | XP_016858235.1 | 8 | 964 | |||
XM_017002747.1 | XP_016858236.1 | 8 | 958 | |||
GenBank mRNA | AF257077.1 | - | 7 | 860 | ||
AK022399.1 | - | 7 | 859 | |||
AK024006.1 | - | 7 | 885 | |||
AK314668.1 | - | 7 | 870 | |||
AL834288.1 | - | 7 | 862 | |||
BC018728.1 | - | 7 | 855 | 4849789 | ||
BC021017.1 | - | 7 | 848 | |||
CR457285.1 | - | 3 | 251 |