Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024074.2 | NP_001019245.1 | 1 | 324 | ||
NM_001024075.1 | NP_001019246.1 | 1 | 324 | |||
NM_006895.2 | NP_008826.1 | 1 | 324 | |||
XM_011511064.2 | XP_011509366.1 | 1 | 140 | |||
XM_017003948.1 | XP_016859437.1 | 1 | 494 | |||
XM_017003949.1 | XP_016859438.1 | 1 | 157 | |||
GenBank mRNA | AF523356.1 | - | 1 | 249 | ||
AF523357.1 | - | 1 | 249 | |||
AF523358.1 | - | 1 | 251 | |||
AF523359.1 | - | 1 | 251 | |||
AF523360.1 | - | 1 | 251 | |||
AK301462.1 | - | 1 | 82 | |||
AK313804.1 | - | 1 | 82 | |||
BC005907.1 | - | 1 | 89 | |||
BC020677.1 | - | 1 | 83 | 4767331 | ||
D16224.1 | - | 1 | 80 | |||
U08092.1 | - | 1 | 109 |