Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024074.2 | NP_001019245.1 | 1 | 282 | ||
NM_001024075.1 | NP_001019246.1 | 1 | 282 | |||
NM_006895.2 | NP_008826.1 | 1 | 282 | |||
XM_011511064.2 | XP_011509366.1 | 1 | 98 | |||
XM_017003948.1 | XP_016859437.1 | 1 | 452 | |||
XM_017003949.1 | XP_016859438.1 | 1 | 115 | |||
GenBank mRNA | AF523356.1 | - | 1 | 207 | ||
AF523357.1 | - | 1 | 207 | |||
AF523358.1 | - | 1 | 209 | |||
AF523359.1 | - | 1 | 209 | |||
AF523360.1 | - | 1 | 209 | |||
AK301462.1 | - | 1 | 40 | |||
AK313804.1 | - | 1 | 40 | |||
BC005907.1 | - | 1 | 47 | |||
BC020677.1 | - | 1 | 41 | 4767331 | ||
D16224.1 | - | 1 | 38 | |||
U08092.1 | - | 1 | 67 |