Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002159.3 | NP_002150.1 | 2 | 127 | ||
GenBank mRNA | BC017835.1 | - | 2 | 100 | 4289874 | |
BP335073.2 | - | 2 | 104 | |||
CB984538.1 | - | 2 | 131 | |||
CR627329.1 | - | 1 | 229 | |||
M26664.1 | - | 1 | 42 |