Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123066.3 | NP_001116538.2 | 15 | 4509 | ||
NM_001123067.3 | NP_001116539.1 | 11 | 3417 | |||
NM_001203251.1 | NP_001190180.1 | 10 | 3324 | |||
NM_001203252.1 | NP_001190181.1 | 11 | 3411 | |||
NM_005910.5 | NP_005901.2 | 12 | 3504 | |||
NM_016834.4 | NP_058518.1 | 10 | 3330 | |||
NM_016835.4 | NP_058519.3 | 14 | 4455 | |||
NM_016841.4 | NP_058525.1 | 9 | 3237 | |||
XM_005257362.4 | XP_005257419.1 | 14 | 4547 | |||
XM_005257364.4 | XP_005257421.1 | 13 | 4460 | |||
XM_005257365.4 | XP_005257422.1 | 13 | 4454 | |||
XM_005257366.3 | XP_005257423.1 | 12 | 4415 | |||
XM_005257367.4 | XP_005257424.1 | 13 | 4349 | |||
XM_005257368.4 | XP_005257425.1 | 12 | 4256 | |||
XM_005257369.4 | XP_005257426.1 | 13 | 3482 | |||
XM_005257370.4 | XP_005257427.1 | 12 | 3459 | |||
XM_005257371.4 | XP_005257428.1 | 11 | 3308 | |||
GenBank mRNA | AB073354.1 | - | 1 | 1802 | ||
AK055986.1 | - | 1 | 642 | |||
AK095802.1 | - | 10 | 3066 | |||
BC094805.1 | - | 1 | 958 |