Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 1964 | ||
GenBank mRNA | AB449912.1 | - | 6 | 1853 | ||
AK293521.1 | - | 5 | 1794 | |||
AK293875.1 | - | 6 | 1923 | |||
AK294776.1 | - | 6 | 1862 | |||
AK297924.1 | - | 3 | 1369 | |||
AK299561.1 | - | 1 | 892 | |||
AK316199.1 | - | 4 | 1644 | |||
BC026050.1 | - | 1 | 887 | |||
BC036942.1 | - | 3 | 1393 | |||
BC052609.1 | - | 5 | 1754 | |||
BC082255.1 | - | 4 | 1864 | |||
BC101817.1 | - | 6 | 1890 | 8069323 | ||
BC101819.1 | - | 6 | 1890 | 8069325 | ||
BC125088.1 | - | 6 | 1879 | 40117866 | ||
BC125089.1 | - | 5 | 1724 | |||
BG328213.1 | - | 1 | 75 | |||
L08240.1 | - | 5 | 1743 |