Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 5 | 1217 | ||
XM_011527126.1 | XP_011525428.1 | 4 | 938 | |||
GenBank mRNA | BC005924.1 | - | 5 | 1182 | 4043628 | |
M23575.1 | - | 5 | 1316 | |||
M27907.1 | - | 2 | 301 | |||
M34420.1 | - | 5 | 1166 | |||
M37399.1 | - | 5 | 1171 | |||
M93061.1 | - | 5 | 1143 | |||
S59493.1 | - | 5 | 1143 |