Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031850.3 | NP_001027020.1 | 4 | 919 | ||
NM_002782.4 | NP_002773.1 | 4 | 919 | |||
GenBank mRNA | BC020652.1 | - | 4 | 886 | 4770562 | |
M31125.1 | - | 4 | 832 | |||
M33666.1 | - | 4 | 839 | |||
X17098.1 | - | 4 | 840 |