Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031850.3 | NP_001027020.1 | 2 | 548 | ||
NM_002782.4 | NP_002773.1 | 2 | 548 | |||
GenBank mRNA | BC020652.1 | - | 2 | 515 | 4770562 | |
DA833634.1 | - | 2 | 548 | |||
M31125.1 | - | 2 | 461 | |||
M33666.1 | - | 2 | 468 | |||
X17098.1 | - | 2 | 469 |