Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256678.1 | NP_001243607.1 | 3 | 1707 | ||
NM_001256679.1 | NP_001243608.1 | 4 | 1202 | |||
NM_001256680.1 | NP_001243609.1 | 3 | 519 | |||
NM_001256681.1 | NP_001243610.1 | 2 | 471 | |||
NM_014672.3 | NP_055487.2 | 4 | 1755 | |||
XM_005268237.3 | XP_005268294.1 | 4 | 1654 | |||
XM_011537409.2 | XP_011535711.1 | 4 | 1581 | |||
XM_011537410.2 | XP_011535712.1 | 4 | 1639 | |||
XM_017021835.1 | XP_016877324.1 | 4 | 1527 | |||
XM_017021836.1 | XP_016877325.1 | 3 | 1591 | |||
XM_017021837.1 | XP_016877326.1 | 3 | 1590 | |||
GenBank mRNA | AB002389.1 | - | 3 | 1457 | ||
AK301931.1 | - | 4 | 976 | |||
AK303508.1 | - | 3 | 318 | |||
AK304066.1 | - | 3 | 1464 | |||
BC032221.1 | - | 4 | 1506 | 5206545 | ||
BC044580.1 | - | 4 | 1524 | 4822711 | ||
BQ063741.1 | - | 2 | 335 | |||
BX161394.1 | - | 4 | 1487 | |||
BX161487.1 | - | 3 | 349 | |||
DC404874.1 | - | 3 | 519 |