Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032279.1 | NP_001027450.1 | 3, 4 | 416 | ||
NM_005133.2 | NP_005124.1 | 3, 4 | 420 | |||
GenBank mRNA | AF121951.1 | - | 3, 4 | 420 | ||
AL713740.1 | - | 2, 3 | 1115 | |||
BC052622.1 | - | 3, 4 | 377 | |||
BC093726.1 | - | 3, 4 | 416 | 7939571 | ||
BC093728.1 | - | 3, 4 | 415 | 7939573 | ||
Y13835.2 | - | 3, 4 | 389 |