Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018108.3 | NP_001018118.1 | 3 | 678 | ||
NM_001199875.1 | NP_001186804.1 | 3 | 678 | |||
NM_001199876.1 | NP_001186805.1 | 3 | 567 | |||
NM_001199877.1 | NP_001186806.1 | 5 | 1076 | |||
NM_001199878.1 | NP_001186807.1 | 3 | 371 | |||
NR_037672.1 | 3 | 544 | ||||
GenBank mRNA | AF073298.1 | - | 3 | 202 | ||
AF320073.1 | - | 3 | 196 | |||
AK000479.1 | - | 3 | 190 | |||
AK307547.1 | - | 3 | 324 | |||
AL049976.1 | - | 3 | 215 | |||
BC008214.1 | - | 3 | 678 | |||
BC009869.2 | - | 5 | 1074 | |||
BC015491.1 | - | 3 | 175 | 3902770 | ||
BC022326.1 | - | 3 | 193 | 4134511 | ||
BC038893.1 | - | 3 | 159 | |||
BI260545.1 | - | 3 | 278 | |||
BP317864.2 | - | 3 | 221 | |||
DA302996.1 | - | 3 | 372 | |||
DB060650.1 | - | 3 | 200 |