Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 5 | 610 | ||
NM_033160.6 | NP_149350.3 | 5 | 453 | |||
NR_134255.1 | 4 | 498 | ||||
XM_005272515.4 | XP_005272572.1 | 5 | 560 | |||
XM_011545679.2 | XP_011543981.1 | 5 | 384 | |||
XM_017014614.1 | XP_016870103.1 | 4 | 2746 | |||
GenBank mRNA | AK057375.1 | - | 5 | 400 | ||
AK292150.1 | - | 5 | 592 | |||
BC031626.1 | - | 5 | 534 | 5172270 | ||
DA103495.1 | - | 5 | 454 | |||
DA120232.1 | - | 5 | 392 |
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 5 | 449 | |||
GenBank mRNA | DA115965.1 | - | 5 | 445 | ||
DR001461.1 | - | 5 | 439 |