Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317916.1 | NP_001304845.1 | 5 | 711 | ||
NM_033160.6 | NP_149350.3 | 5 | 554 | |||
NR_134255.1 | 4 | 599 | ||||
XM_005272515.4 | XP_005272572.1 | 5 | 661 | |||
XM_011545679.2 | XP_011543981.1 | 5 | 485 | |||
XM_017014614.1 | XP_016870103.1 | 4 | 2847 | |||
GenBank mRNA | AK057375.1 | - | 5 | 501 | ||
AK292150.1 | - | 5 | 693 | |||
BC031626.1 | - | 5 | 635 | 5172270 | ||
DA103495.1 | - | 5 | 555 | |||
DA120232.1 | - | 5 | 493 | |||
DA404383.1 | - | 5 | 551 |
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 5 | 550 | |||
GenBank mRNA | DA115965.1 | - | 5 | 546 | ||
DR001461.1 | - | 5 | 540 |