Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF658B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003528.3 | 4, 5 | 385 | |||
GenBank mRNA | DA115965.1 | - | 4, 5 | 381 | ||
DR001461.1 | - | 4, 5 | 375 |
Gene Symbol: ZNF658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057375.1 | - | 4, 5 | 336 | ||
AK292150.1 | - | 4, 5 | 528 | |||
BC031626.1 | - | 4, 5 | 470 | 5172270 | ||
DA103495.1 | - | 4, 5 | 390 | |||
DA404383.1 | - | 4, 5 | 386 |