Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164803.1 | NP_001158275.1 | 5, 6 | 665 | ||
NM_002139.3 | NP_002130.2 | 7, 8 | 990 | |||
NR_028476.1 | 6, 7 | 818 | ||||
NR_028477.1 | 7, 8 | 1025 | ||||
GenBank mRNA | AK091520.1 | - | 6, 7 | 764 | ||
AK096015.1 | - | 2 | 896 | |||
AK304573.1 | - | 7, 8 | 970 | |||
AK304868.1 | - | 5, 6 | 610 | |||
AK309911.1 | - | 7, 8 | 945 | |||
AY464692.1 | - | 6, 7 | 780 | |||
BC006550.2 | - | 7, 8 | 926 | 3010123 | ||
BC007435.2 | - | 7, 8 | 926 | 3010123 | ||
BX647131.1 | - | 6, 7 | 761 |