Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278205.1 | NP_001265134.1 | 2, 3 | 383 | ||
NM_016299.3 | NP_057383.2 | 2, 3 | 383 | |||
NR_103464.1 | 2, 3 | 383 | ||||
GenBank mRNA | AF112210.1 | - | 2, 3 | 202 | ||
AF143723.1 | - | 2, 3 | 192 | |||
AK000880.1 | - | 2, 3 | 224 | |||
AK225265.1 | - | 2, 3 | 227 | |||
AK225651.1 | - | 2, 3 | 227 | |||
AK292323.1 | - | 2, 3 | 191 | |||
AK302452.1 | - | 2, 3 | 227 | |||
BC065281.1 | - | 2, 3 | 214 | |||
BC108738.1 | - | 2, 3 | 317 | |||
BC119690.1 | - | 2, 3 | 202 | 40116697 | ||
BP201211.1 | - | 2, 3 | 383 | |||
BQ424016.1 | - | 2, 3 | 314 | |||
CN308284.1 | - | 2, 3 | 220 |