Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017820.4 | NP_060290.3 | 22 | 2836 | ||
XM_011518810.1 | XP_011517112.1 | 24 | 3301 | |||
XM_011518814.1 | XP_011517116.1 | 16 | 2251 | |||
XM_011518815.1 | XP_011517117.1 | 13 | 1935 | |||
XM_017014861.1 | XP_016870350.1 | 25 | 3445 | |||
XM_017014862.1 | XP_016870351.1 | 23 | 3186 | |||
XM_017014863.1 | XP_016870352.1 | 23 | 3144 | |||
XM_017014864.1 | XP_016870353.1 | 22 | 3506 | |||
XM_017014865.1 | XP_016870354.1 | 22 | 3153 | |||
XM_017014866.1 | XP_016870355.1 | 21 | 2809 | |||
XM_017014867.1 | XP_016870356.1 | 21 | 2850 | |||
GenBank mRNA | AK055004.1 | - | 12 | 1968 | ||
AK094438.1 | - | 22 | 2736 | |||
BU947354.1 | - | 4 | 558 |