Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037175.2 | NP_001032252.1 | 4 | 615 | ||
NM_017982.3 | NP_060452.3 | 4 | 615 | |||
XM_005273169.1 | XP_005273226.1 | 5 | 632 | |||
XM_005273172.1 | XP_005273229.1 | 5 | 695 | |||
XM_006711408.1 | XP_006711471.1 | 4 | 613 | |||
XM_011509685.1 | XP_011507987.1 | 4 | 647 | |||
XM_011509687.1 | XP_011507989.1 | 5 | 695 | |||
XM_017001583.1 | XP_016857072.1 | 5 | 813 | |||
XM_017001584.1 | XP_016857073.1 | 5 | 812 | |||
XM_017001585.1 | XP_016857074.1 | 6 | 796 | |||
XM_017001586.1 | XP_016857075.1 | 5 | 690 | |||
XM_017001587.1 | XP_016857076.1 | 5 | 632 | |||
XM_017001588.1 | XP_016857077.1 | 4 | 647 | |||
XM_017001589.1 | XP_016857078.1 | 5 | 817 | |||
GenBank mRNA | AK000914.1 | - | 4 | 496 | ||
AK096265.1 | - | 3 | 1069 | |||
AK295553.1 | - | 3 | 365 | |||
AK296923.1 | - | 5 | 695 | |||
AY358495.1 | - | 4 | 592 | |||
BC004888.2 | - | 4 | 496 | 3926397 | ||
EF560748.1 | - | 4 | 543 |