Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020207.4 | NP_064592.2 | 18 | 4029 | ||
XM_011518641.2 | XP_011516943.1 | 17 | 3876 | |||
XM_011518644.2 | XP_011516946.1 | 17 | 3821 | |||
XM_011518645.2 | XP_011516947.1 | 18 | 4029 | |||
XM_011518648.2 | XP_011516950.1 | 9 | 2101 | |||
XM_011518650.2 | XP_011516952.1 | 18 | 3348 | |||
XM_017014707.1 | XP_016870196.1 | 18 | 4016 | |||
GenBank mRNA | AK308131.1 | - | 3 | 1064 | ||
AL133663.1 | - | 13 | 2629 | |||
BC022957.1 | - | 3 | 458 | 4995030 | ||
BG108141.1 | - | 2 | 289 |