Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012288.2 | NP_001012288.2 | 4 | 501 | ||
NM_022148.3 | NP_071431.2 | 5 | 604 | |||
XM_011546181.2 | XP_011544483.1 | 5 | 639 | |||
GenBank mRNA | AB052639.1 | - | 5 | 515 | ||
AF142570.1 | - | 5 | 531 | |||
AF338733.1 | - | 5 | 515 | |||
AK223303.1 | - | 5 | 631 | |||
AY775789.1 | - | 4 | 412 |