Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032341.4 | NP_115717.3 | 4 | 818 | ||
GenBank mRNA | AK057374.1 | - | 2 | 260 | ||
AK293006.1 | - | 4 | 818 | |||
BC006011.1 | - | 4 | 733 | 4296688 |