Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 8 | 1128 | ||
XM_005273603.4 | XP_005273660.1 | 7 | 1101 | |||
XM_005273604.4 | XP_005273661.1 | 6 | 1020 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 921 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 959 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1122 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1173 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1140 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1042 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1103 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1040 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1022 | |||
GenBank mRNA | AK297741.1 | - | 5 | 703 | ||
AK298940.1 | - | 4 | 821 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1071 | ||
NR_003494.1 | 6 | 805 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1166 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1236 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 980 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 823 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1245 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1184 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1212 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1133 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1105 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 694 | |||
GenBank mRNA | AK098138.1 | - | 6 | 772 | ||
AK308439.1 | - | 7 | 886 | |||
AL529666.3 | - | 5 | 742 | |||
BC062615.1 | - | 6 | 849 | |||
BC085016.1 | - | 7 | 878 | |||
BC098427.1 | - | 6 | 855 |