Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027786.1 | 9, 10 | 800 | |||
GenBank mRNA | AK094872.1 | - | 8, 9 | 1156 | ||
AL365513.1 | - | 9, 10 | 789 |