Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138422.2 | NP_612431.2 | 2 | 1001 | ||
GenBank mRNA | BC011824.1 | - | 2 | 955 | 3830276 |