Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271844.1 | NP_001258773.1 | 6, 7 | 889 | ||
NM_001271845.1 | NP_001258774.1 | 6, 7 | 928 | |||
NM_019020.3 | NP_061893.2 | 10, 11 | 2044 | |||
XM_005257049.2 | XP_005257106.1 | 10, 11 | 2055 | |||
XM_005257050.3 | XP_005257107.1 | 10, 11 | 2058 | |||
XM_006721694.3 | XP_006721757.1 | 10, 11 | 2061 | |||
XM_011524322.2 | XP_011522624.1 | 10, 11 | 2019 | |||
XM_011524323.2 | XP_011522625.2 | 8, 9 | 1969 | |||
XM_011524324.2 | XP_011522626.1 | 6, 7 | 1513 | |||
XM_017024186.1 | XP_016879675.1 | 8, 9 | 1796 | |||
XM_017024187.1 | XP_016879676.1 | 9, 10 | 2036 | |||
XM_017024188.1 | XP_016879677.1 | 8, 9 | 1176 | |||
GenBank mRNA | AB449902.1 | - | 9, 10 | 1918 | ||
BC028290.1 | - | 10, 11 | 2034 | 4480654 | ||
BC036947.1 | - | 6, 7 | 933 | 5392579 | ||
CR936670.1 | - | 6, 7 | 894 |