Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182497.3 | NP_872303.2 | 4 | 658 | ||
XM_011524339.2 | XP_011522641.2 | 2 | 2425 | |||
XM_017024189.1 | XP_016879678.1 | 2 | 2425 | |||
GenBank mRNA | AJ786658.1 | - | 2 | 498 | ||
AK093919.1 | - | 4 | 659 | |||
BC130396.1 | - | 3 | 533 | 40146427 | ||
BC144250.1 | - | 3 | 533 | 9052770 | ||
BC150174.1 | - | 4 | 658 | 40147687 | ||
BC150191.1 | - | 4 | 658 | 40148622 |