Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011717.1 | NP_001011717.1 | 9 | 434 | ||
GenBank mRNA | AB164464.1 | - | 9 | 434 |