Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM101A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181709.4 | NP_859060.3 | 3 | 1471 | ||
GenBank mRNA | AK074545.1 | - | 7 | 1249 | ||
AK126577.1 | - | 3 | 1471 | |||
BC042102.1 | - | 3 | 1378 | |||
BC085606.1 | - | 1 | 225 | |||
BC141805.1 | - | 2 | 1239 | 40147249 |
Gene Symbol: ZNF664-FAM101A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204299.1 | NP_001191228.1 | 5 | 1734 |