Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT16P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002949.2 | 3 | 1500 | |||
NR_027766.1 | 2 | 1574 | ||||
GenBank mRNA | AK000532.1 | - | 3 | 1451 | ||
BC015593.2 | - | 2 | 1536 |